COMH4211AFCD Combi-Blocks, Inc.

tert-Butyl 4-bromo-2-(trifluoromethyl)benzoate

  • MDL number MFCD28406073
  • Cas Number : 1416979-56-0
  • Molecular Weight : 325.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3149
Availability details for
Quantity
  • Availability
  • Add to Cart