COMH4211AFCE Combi-Blocks, Inc.

5-Hydroxy-2-(2-methoxy-ethoxy)-n,n-dimethyl-benzamide

  • MDL number MFCD31618918
  • Cas Number : 2270908-58-0
  • Molecular Weight : 239.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3151

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3151
Availability details for
Quantity
  • Availability
  • Add to Cart