COMH4211AFCF Combi-Blocks, Inc.

Methyl 5-bromo-2-(methylsulfanyl)pyridine-3-carboxylate

  • MDL number MFCD31540233
  • Cas Number : 1220422-12-7
  • Molecular Weight : 262.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-bromo-2-(methylthio)nicotinate
Inchi Code 1S/C8H8BrNO2S/c1-12-8(11)6-3-5(9)4-10-7(6)13-2/h3-4H,1-2H3
Inchi Key FFKOQOWUJKHKAM-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3153

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3153
Availability details for
Quantity
  • Availability
  • Add to Cart