COMH4211AFD0 Combi-Blocks, Inc.

4-Nitropiperidine

  • MDL number MFCD13178717
  • Cas Number : 890853-07-3
  • Molecular Weight : 130.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3154

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3154
Availability details for
Quantity
  • Availability
  • Add to Cart