COMH4211AFD1 Combi-Blocks, Inc.

(2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(4-methylpiperazin-1-yl)methanone

  • MDL number MFCD28402149
  • Cas Number : 1092563-26-2
  • Molecular Weight : 344.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3156

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3156
Availability details for
Quantity
  • Availability
  • Add to Cart