COMH4211AFD3 Combi-Blocks, Inc.

4-Bromo-1-cyclobutyl-1h-indole-3-carbonitrile

  • MDL number MFCD31583015
  • Cas Number : 2270906-90-4
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3159
Availability details for
Quantity
  • Availability
  • Add to Cart