COMH4211AFD4 Combi-Blocks, Inc.

5-Bromo-2-(3-methoxyphenyl)pyridine

  • MDL number MFCD14707259
  • Cas Number : 1119089-23-4
  • Molecular Weight : 264.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2-(3-methoxyphenyl)pyridine
Inchi Code 1S/C12H10BrNO/c1-15-11-4-2-3-9(7-11)12-6-5-10(13)8-14-12/h2-8H,1H3
Inchi Key DTTWWGLYRMBSGR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3163
Availability details for
Quantity
  • Availability
  • Add to Cart