COMH4211AFD6 Combi-Blocks, Inc.

1-(4-Ethoxyphenyl)cyclopropanecarbonitrile

  • MDL number MFCD00297176
  • Cas Number : 74205-06-4
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3167

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3167
Availability details for
Quantity
  • Availability
  • Add to Cart