COMH4211AFDB Combi-Blocks, Inc.

5-Bromo-1-(2-methoxy-ethyl)-1h-benzimidazole

  • MDL number MFCD31579428
  • Cas Number : 1374145-25-1
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3185

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3185
Availability details for
Quantity
  • Availability
  • Add to Cart