COMH4211AFDE Combi-Blocks, Inc.

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,4-dihydro-2h-benzo[d][1,3]oxazin-2-one

  • MDL number MFCD30829394
  • Cas Number : 1899832-38-2
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3195

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3195
Availability details for
Quantity
  • Availability
  • Add to Cart