COMH4211AFE2 Combi-Blocks, Inc.

4-(5-Bromo-2-nitro-phenoxymethyl)-tetrahydro-pyran

  • MDL number MFCD30729447
  • Cas Number : 2205384-90-1
  • Molecular Weight : 316.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3207

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3207
Availability details for
Quantity
  • Availability
  • Add to Cart