COMH4211AFE3 Combi-Blocks, Inc.

4-Bromo-1-(methoxymethoxy)-2-nitrobenzene

  • MDL number MFCD31381615
  • Cas Number : 1396779-75-1
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3210

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3210
Availability details for
Quantity
  • Availability
  • Add to Cart