COMH4211AFE4 Combi-Blocks, Inc.

4-Methyl-3-(propan-2-yloxy)aniline

  • MDL number MFCD11934532
  • Cas Number : 111185-05-8
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3213

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3213
Availability details for
Quantity
  • Availability
  • Add to Cart