COMH4211AFE5 Combi-Blocks, Inc.

1-4-[(Ethylamino)carbonyl]phenyl-5-isopropyl-1h-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD31613690
  • Cas Number : 912839-06-6
  • Molecular Weight : 302.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3215

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3215
Availability details for
Quantity
  • Availability
  • Add to Cart