COMH4211AFE6 Combi-Blocks, Inc.

4-(Methoxymethyl)bicyclo[2.2.2]octane-1-carboxylic acid

  • MDL number MFCD31381610
  • Cas Number : 2168336-36-3
  • Molecular Weight : 198.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3218

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3218
Availability details for
Quantity
  • Availability
  • Add to Cart