COMH4211AFE7 Combi-Blocks, Inc.

6-Bromo-1-(vinylsulfonyl)indoline

  • MDL number MFCD31579421
  • Cas Number : 1912446-80-0
  • Molecular Weight : 288.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3220

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3220
Availability details for
Quantity
  • Availability
  • Add to Cart