COMH4211AFE9 Combi-Blocks, Inc.

5-Bromo-2-(cyclopropylmethoxy)aniline

  • MDL number MFCD11136592
  • Cas Number : 1019590-24-9
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3224
Availability details for
Quantity
  • Availability
  • Add to Cart