COMH4211AFEB Combi-Blocks, Inc.

5-Hydroxy-2-isopropoxy-benzoic acid methyl ester

  • MDL number MFCD24729546
  • Cas Number : 927908-25-6
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3228
Availability details for
Quantity
  • Availability
  • Add to Cart