COMH4211AFED Combi-Blocks, Inc.

6-(Morpholine-4-carbonyl)-1h-indole-3-carbaldehyde

  • MDL number MFCD31539540
  • Cas Number : 1256353-21-5
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3232
Availability details for
Quantity
  • Availability
  • Add to Cart