COMH4211AFEF Combi-Blocks, Inc.

3-(3-Ethoxy-phenyl)-2-methyl-propionic acid

  • MDL number MFCD09894711
  • Cas Number : 1017369-54-8
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-ethoxyphenyl)-2-methylpropanoic acid
Inchi Code 1S/C12H16O3/c1-3-15-11-6-4-5-10(8-11)7-9(2)12(13)14/h4-6,8-9H,3,7H2,1-2H3,(H,13,14)
Inchi Key LHHHNVSYSLFAJR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3240

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3240
Availability details for
Quantity
  • Availability
  • Add to Cart