COMH4211AFF1 Combi-Blocks, Inc.

6-Bromo-3-iodo-7-methylimidazo[1,2-a]pyridine

  • MDL number MFCD22628058
  • Cas Number : 1246184-61-1
  • Molecular Weight : 337.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3243

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3243
Availability details for
Quantity
  • Availability
  • Add to Cart