COMH4211AFF2 Combi-Blocks, Inc.

4-Bromo-2-(cyclopropylmethoxy)benzoic acid

  • MDL number MFCD11651844
  • Cas Number : 1094474-68-6
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3245

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3245
Availability details for
Quantity
  • Availability
  • Add to Cart