COMH4211AFF3 Combi-Blocks, Inc.

N-(4-Aminophenyl)-2-chloro-4-fluorobenzamide

  • MDL number MFCD09043964
  • Cas Number : 926262-97-7
  • Molecular Weight : 264.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3249

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3249
Availability details for
Quantity
  • Availability
  • Add to Cart