COMH4211AFF7 Combi-Blocks, Inc.

Cis/trans (4-benzyloxycarbonylamino-cyclohexyl)-acetic acid

  • MDL number MFCD31561594
  • Cas Number : 1823505-57-2
  • Molecular Weight : 291.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3255

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3255
Availability details for
Quantity
  • Availability
  • Add to Cart