COMH4211AFF8 Combi-Blocks, Inc.

2-Cyclopentyl-n-methoxy-n-methylacetamide

  • MDL number MFCD18839866
  • Cas Number : 134560-38-6
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3256

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3256
Availability details for
Quantity
  • Availability
  • Add to Cart