COMH4211AFFB Combi-Blocks, Inc.

3-((4-Aminophenyl)thio)dihydrofuran-2(3h)-one

  • MDL number MFCD11635214
  • Cas Number : 1095660-34-6
  • Molecular Weight : 209.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3267

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3267
Availability details for
Quantity
  • Availability
  • Add to Cart