COMH4211AFFC Combi-Blocks, Inc.

3-([(4-Bromo-2-fluorophenyl)methyl]amino)butan-1-ol

  • MDL number MFCD25980717
  • Cas Number : 1553259-05-4
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3268

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3268
Availability details for
Quantity
  • Availability
  • Add to Cart