COMH4211AFFD Combi-Blocks, Inc.

Methyl 6-bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD22373271
  • Cas Number : 1121058-16-9
  • Molecular Weight : 323.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3270
Availability details for
Quantity
  • Availability
  • Add to Cart