COMH4211AFFF Combi-Blocks, Inc.

4-(Cyclopropylmethoxy)-3-methylaniline

  • MDL number MFCD11214171
  • Cas Number : 1094646-64-6
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3274

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3274
Availability details for
Quantity
  • Availability
  • Add to Cart