COMH4211B001 Combi-Blocks, Inc.

Cyclopropylmethyl-(2-trifluoromethoxy-phenyl)-amine

  • MDL number MFCD12138729
  • Cas Number : 1156163-83-5
  • Molecular Weight : 231.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3287

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3287
Availability details for
Quantity
  • Availability
  • Add to Cart