COMH4211B003 Combi-Blocks, Inc.

6-[2-Fluoro-1-(fluoromethyl)ethoxy]nicotinic acid

  • MDL number MFCD24499134
  • Cas Number : 1072855-42-5
  • Molecular Weight : 217.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-((1,3-difluoropropan-2-yl)oxy)nicotinic acid
Inchi Code 1S/C9H9F2NO3/c10-3-7(4-11)15-8-2-1-6(5-12-8)9(13)14/h1-2,5,7H,3-4H2,(H,13,14)
Inchi Key DNLIHJCOUQKUQX-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3289

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3289
Availability details for
Quantity
  • Availability
  • Add to Cart