COMH4211B004 Combi-Blocks, Inc.

3,4-Difluoro-5-methoxybenzonitrile

  • MDL number MFCD22490584
  • Cas Number : 1262552-14-6
  • Molecular Weight : 169.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3290

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3290
Availability details for
Quantity
  • Availability
  • Add to Cart