COMH4211B005 Combi-Blocks, Inc.

(1-Cyclopropanesulfonyl-piperidin-4-yl)-methanol

  • MDL number MFCD20352475
  • Cas Number : 1036738-95-0
  • Molecular Weight : 219.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3291

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3291
Availability details for
Quantity
  • Availability
  • Add to Cart