COMH4211B006 Combi-Blocks, Inc.

2-(3-Trifluoromethoxy-benzyl)-butyric acid

  • MDL number MFCD31583263
  • Cas Number : 2270905-09-2
  • Molecular Weight : 262.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3294

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3294
Availability details for
Quantity
  • Availability
  • Add to Cart