COMH4211B008 Combi-Blocks, Inc.

3-(2-Chloropyridin-4-yloxy)-4-fluorobenzoic acid

  • MDL number MFCD31614091
  • Cas Number : 1268619-81-3
  • Molecular Weight : 267.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3296

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3296
Availability details for
Quantity
  • Availability
  • Add to Cart