COMH4211B009 Combi-Blocks, Inc.

5-Bromo-1-(piperidin-4-ylmethyl)-1h-indole hydrochloride

  • MDL number MFCD31561615
  • Cas Number : 1808957-59-6
  • Molecular Weight : 329.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3297

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3297
Availability details for
Quantity
  • Availability
  • Add to Cart