COMH4211B00A Combi-Blocks, Inc.

3-Bromo-4-(2,4-difluorophenoxy)benzaldehyde

  • MDL number MFCD31614036
  • Cas Number : 183110-68-1
  • Molecular Weight : 313.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3298

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3298
Availability details for
Quantity
  • Availability
  • Add to Cart