COMH4211B00B Combi-Blocks, Inc.

1-(2,2-Difluoro-2-(4-fluorophenyl)ethyl)piperidin-4-amine

  • MDL number MFCD31618976
  • Cas Number : 1961320-13-7
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3299

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3299
Availability details for
Quantity
  • Availability
  • Add to Cart