COMH4211B00E Combi-Blocks, Inc.

[3-(3-Hydroxy-azetidin-1-yl)-propyl]-carbamic acid benzyl ester

  • MDL number MFCD31613930
  • Cas Number : 2270906-23-3
  • Molecular Weight : 264.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3310
Availability details for
Quantity
  • Availability
  • Add to Cart