COMH4211B012 Combi-Blocks, Inc.

1-(4-Cyclopropylcarbamoyl-phenyl)-5-propyl-1h-[1,2,3]triazole-4-carboxylic acid

  • MDL number MFCD31583041
  • Cas Number : 912838-20-1
  • Molecular Weight : 314.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3316

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3316
Availability details for
Quantity
  • Availability
  • Add to Cart