COMH4211B014 Combi-Blocks, Inc.

2,5-Dichloro-n-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]benzenesulfonamide

  • MDL number MFCD28406027
  • Cas Number : 1426214-54-1
  • Molecular Weight : 428.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3322

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3322
Availability details for
Quantity
  • Availability
  • Add to Cart