COMH4211B015 Combi-Blocks, Inc.

5-Bromo-n-ethyl-2-methoxybenzamide

  • MDL number MFCD08104770
  • Cas Number : 899143-59-0
  • Molecular Weight : 258.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3323

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3323
Availability details for
Quantity
  • Availability
  • Add to Cart