COMH4211B016 Combi-Blocks, Inc.

Chloromethyl 2-cyclopentylacetate

  • MDL number MFCD20634816
  • Cas Number : 344326-08-5
  • Molecular Weight : 176.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3326
Availability details for
Quantity
  • Availability
  • Add to Cart