COMH4211B017 Combi-Blocks, Inc.

5-(2-Chlorophenyl)pyrazin-2-amine

  • MDL number MFCD21253696
  • Cas Number : 71553-78-1
  • Molecular Weight : 205.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3329

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3329
Availability details for
Quantity
  • Availability
  • Add to Cart