COMH4211B018 Combi-Blocks, Inc.

4-Allyloxy-1-benzyl-1h-pyrazole

  • MDL number MFCD31561506
  • Cas Number : 1326316-19-1
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3332

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3332
Availability details for
Quantity
  • Availability
  • Add to Cart