COMH4211B01B Combi-Blocks, Inc.

6-(Cyclobutyloxy)nicotinic acid

  • MDL number MFCD16840930
  • Cas Number : 1072855-44-7
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3336

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3336
Availability details for
Quantity
  • Availability
  • Add to Cart