COMH4211B01C Combi-Blocks, Inc.

(2S)-1-(Cyclopropanesulfonyl)-2-methylpiperazine

  • MDL number MFCD24301033
  • Cas Number : 1568016-77-2
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3340
Availability details for
Quantity
  • Availability
  • Add to Cart