COMH4211B01D Combi-Blocks, Inc.

3-Bromo-4-(2,4-difluorobenzyloxy)-1h-pyridin-2-one

  • MDL number MFCD25965052
  • Cas Number : 586373-59-3
  • Molecular Weight : 316.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3342

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3342
Availability details for
Quantity
  • Availability
  • Add to Cart