COMH4211B01F Combi-Blocks, Inc.

1-(3-Bromophenyl)butan-1-one

  • MDL number MFCD12153327
  • Cas Number : 207994-95-4
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3349

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3349
Availability details for
Quantity
  • Availability
  • Add to Cart