COMH4211B020 Combi-Blocks, Inc.

2,5-Dioxopyrrolidin-1-yl hept-6-ynoate

  • MDL number MFCD29041721
  • Cas Number : 917222-23-2
  • Molecular Weight : 223.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3351

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3351
Availability details for
Quantity
  • Availability
  • Add to Cart